Assay Details
| Gene Symbol: SLC39A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139177.3 | NP_631916.2 | 5 - 6 | 539 | 83 | |
| XM_005257134.3 | XP_005257191.1 | 5 - 6 | 626 | 83 | ||
| XM_017024330.1 | XP_016879819.1 | 6 - 7 | 830 | 83 | ||
| XM_017024331.1 | XP_016879820.1 | 6 - 7 | 921 | 83 | ||
| XM_017024332.1 | XP_016879821.1 | 5 - 6 | 536 | 83 | ||
| XM_017024339.1 | XP_016879828.1 | 5 - 6 | 539 | 83 | ||
| XM_017024340.1 | XP_016879829.1 | 5 - 6 | 539 | 83 | ||
| XM_017024341.1 | XP_016879830.1 | 5 - 6 | 626 | 83 | ||
| XM_017024342.1 | XP_016879831.1 | 5 - 6 | 539 | 83 | ||
| GenBank mRNA | AF331643.1 | - | 5 - 6 | 501 | 83 | |
| AK313376.1 | - | 5 - 6 | 512 | 83 | ||
| BC035631.1 | - | 5 - 6 | 518 | 5219031 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SLC39A11
Genomic Map