Assay Details
Gene Symbol: ST3GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006927.3 | NP_008858.1 | 3 - 4 | 1679 | 70 | |
GenBank mRNA | AK127322.1 | - | 2 - 3 | 2644 | 70 | |
AK313020.1 | - | 2 - 3 | 810 | 70 | ||
BC036777.1 | - | 2 - 3 | 578 | 5750330 | 70 | |
U63090.1 | - | 2 - 3 | 1216 | 70 | ||
X96667.1 | - | 2 - 3 | 832 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001236.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
amino sugar metabolic process
cellular protein modification process
oligosaccharide metabolic process
O-glycan processing
keratan sulfate biosynthetic process
protein N-linked glycosylation via asparagine
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
ganglioside biosynthetic process
amino sugar metabolic process
cellular protein modification process
oligosaccharide metabolic process
O-glycan processing
keratan sulfate biosynthetic process
protein N-linked glycosylation via asparagine
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16017062.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017062.hg.1 | ENST00000393640.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16017062.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16017062.hg.1 | TC16001236.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000561708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000561708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000561708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600183148.hg.1 | TC1600010756.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000342907.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000393640.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000561708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000561708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000561708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000566097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | ENST00000567822.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010756.hg.1 | NM_006927.3 | Refseq |
