Assay Details
| Gene Symbol: DDA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024050.5 | NP_076955.1 | 2 - 3 | 212 | 97 | |
| GenBank mRNA | AY563006.2 | - | 2 - 3 | 206 | 97 | |
| BC000615.2 | - | 2 - 3 | 187 | 3346055 | 97 | |
| DQ090952.1 | - | 2 - 3 | 196 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000332.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19004346.hg.1 | ENST00000359866.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004346.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004346.hg.1 | TC19000332.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000594501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000594501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000594501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000594501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000594501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000594501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000594501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000594501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007320.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | NM_024050.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | TC1900007320.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | ENST00000359866.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | ENST00000593466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | ENST00000596582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | ENST00000596925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153227.hg.1 | NM_024050.5 | Refseq |

DDA1
Genomic Map