Assay Details
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006675.4 | NP_006666.1 | 2 - 3 | 164 | 59 | |
XM_011520912.2 | XP_011519214.1 | 2 - 3 | 142 | 59 | ||
GenBank mRNA | AF089749.1 | - | 2 - 3 | 107 | 59 | |
AK027366.1 | - | 2 - 3 | 140 | 59 | ||
AK223179.1 | - | 2 - 3 | 136 | 59 | ||
BC071881.1 | - | 1 - 2 | 79 | 6256950 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000036.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF089749.1,AK027366.1,BC071881.1,NM_006675.4,XM_011520912.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000496.hg.1 | ENST00000011898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000496.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000496.hg.1 | ENST00000537971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000496.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000496.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000496.hg.1 | TC12000036.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12000492.hg.1 | ENST00000011898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000492.hg.1 | ENST00000537971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000492.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000492.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000492.hg.1 | TC12000036.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12000494.hg.1 | ENST00000011898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000494.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000494.hg.1 | TC12000036.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144901.hg.1 | TC1200006520.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144897.hg.1 | TC1200006520.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144894.hg.1 | TC1200006520.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
