Assay Details
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168320.1 | NP_001161792.1 | 3 - 4 | 369 | 62 | |
NM_006675.4 | NP_006666.1 | 4 - 5 | 436 | 62 | ||
XM_011520912.2 | XP_011519214.1 | 5 - 6 | 553 | 62 | ||
XM_011520913.2 | XP_011519215.1 | 2 - 3 | 295 | 62 | ||
GenBank mRNA | AF089749.1 | - | 4 - 5 | 379 | 62 | |
AK027366.1 | - | 4 - 5 | 412 | 62 | ||
AK223179.1 | - | 4 - 5 | 408 | 62 | ||
BC071881.1 | - | 3 - 4 | 351 | 6256950 | 62 | |
CR542109.1 | - | 2 - 3 | 253 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000036.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000499.hg.1 | ENST00000011898.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000499.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000499.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000499.hg.1 | ENST00000537971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12000499.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000499.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12000499.hg.1 | TC12000036.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200144907.hg.1 | TC1200006520.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000011898.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000407263.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000431374.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000444315.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000492305.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000537971.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | ENST00000539631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_001168320.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006520.hg.1 | NM_006675.4 | Refseq |
