Assay Details
| Gene Symbol: DUSP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142314.1 | NP_001135786.1 | 2 - 3 | 649 | 70 | |
| GenBank mRNA | AB063187.1 | - | 2 - 3 | 654 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001092.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein kinase activity
activation of protein kinase activity
peptidyl-tyrosine dephosphorylation
positive regulation of MAPK cascade
positive regulation of JUN kinase activity
negative regulation of JUN kinase activity
positive regulation of protein kinase activity
negative regulation of JNK cascade
positive regulation of JNK cascade
protein tyrosine phosphatase activity
protein kinase inhibitor activity
MAP-kinase scaffold activity
protein binding
JUN kinase phosphatase activity
protein kinase activator activity
mitogen-activated protein kinase kinase kinase binding
negative regulation of protein kinase activity
activation of protein kinase activity
peptidyl-tyrosine dephosphorylation
positive regulation of MAPK cascade
positive regulation of JUN kinase activity
negative regulation of JUN kinase activity
positive regulation of protein kinase activity
negative regulation of JNK cascade
positive regulation of JNK cascade
protein tyrosine phosphatase activity
protein kinase inhibitor activity
MAP-kinase scaffold activity
protein binding
JUN kinase phosphatase activity
protein kinase activator activity
mitogen-activated protein kinase kinase kinase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015141.hg.1 | ENST00000413954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015141.hg.1 | ENST00000413954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016332.hg.1 | TC02001092.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016332.hg.1 | ENST00000342619.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016332.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016332.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016332.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02016332.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010182.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015141.hg.1 | ENST00000413954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015141.hg.1 | ENST00000413954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015141.hg.1 | ENST00000444562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | ENST00000342619.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | ENST00000354221.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | ENST00000469344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | NM_001142314.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | NM_080876.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200171614.hg.1 | TC0200010182.hg.1 | Clariom D TC |

DUSP19
Genomic Map