Assay Details
| Gene Symbol: PIK3CD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001477.1 | XP_016856966.1 | 2 - 3 | 113 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017001477.1 transcripts.
Set Membership: |
|

PIK3CD
Genomic Map