Assay Details
Gene Symbol: VPS13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015378.3 | NP_056193.2 | 59 - 60 | 11642 | 86 | |
NM_018156.3 | NP_060626.2 | 58 - 59 | 11567 | 86 | ||
GenBank mRNA | AB007922.3 | - | 41 - 42 | 7949 | 86 | |
AJ608774.1 | - | 59 - 60 | 11614 | 86 | ||
AJ608775.1 | - | 58 - 59 | 11539 | 86 | ||
AL833079.1 | - | 14 - 15 | 2253 | 86 | ||
AL833320.1 | - | 1968 | 86 | |||
BC028115.1 | - | 17 - 18 | 2558 | 86 | ||
BC051804.1 | - | 7 - 8 | 1044 | 86 | ||
BX641035.1 | - | 33 - 34 | 5130 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000156.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
