Assay Details
Gene Symbol: MED21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073463.1 | 1 - 2 | 101 | 123 | ||
GenBank mRNA | AK309064.1 | - | 1 - 2 | 73 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
blastocyst development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
protein ubiquitination
stem cell population maintenance
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription cofactor activity
transcription coactivator activity
DNA-directed RNA polymerase activity
protein binding
ubiquitin protein ligase activity
blastocyst development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
protein ubiquitination
stem cell population maintenance
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription cofactor activity
transcription coactivator activity
DNA-directed RNA polymerase activity
protein binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | ENST00000621885.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | ENST00000621885.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012612.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | ENST00000282892.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | ENST00000536503.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | ENST00000536711.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | ENST00000546323.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | NM_001271811.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | NM_004264.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | NR_073463.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | NR_073464.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200199595.hg.1 | TC1200012612.hg.1 | Clariom D TC |
