Assay Details
| Gene Symbol: IPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024658.3 | NP_078934.3 | 3 - 4 | 365 | 147 | |
| NR_051979.1 | 3 - 4 | 365 | 147 | |||
| GenBank mRNA | AB208897.1 | - | 3 - 4 | 255 | 147 | |
| AF411122.1 | - | 3 - 4 | 236 | 147 | ||
| AK056527.1 | - | 3 - 4 | 269 | 147 | ||
| AK056702.1 | - | 3 - 4 | 212 | 115 | ||
| AK057044.1 | - | 2 - 3 | 323 | 147 | ||
| AK057277.1 | - | 3 - 4 | 269 | 147 | ||
| AK094897.1 | - | 2 - 3 | 372 | 275 | ||
| AK302097.1 | - | 3 - 4 | 255 | 147 | ||
| AK314305.1 | - | 3 - 4 | 244 | 147 | ||
| BC110804.1 | - | 3 - 4 | 267 | 6164334 | 147 | |
| BC136759.1 | - | 3 - 4 | 316 | 9020749 | 147 | |
| BC144399.1 | - | 3 - 4 | 316 | 147 | ||
| BG333193.1 | - | 3 - 4 | 365 | 147 | ||
| BX248267.1 | - | 3 - 4 | 247 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK056702.1, AK057044.1, AK094897.1, AK302097.1 transcripts.
Set Membership: |
|

IPO4
Genomic Map