Assay Details
Gene Symbol: OBP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110242.1 | 4 - 5 | 436 | 70 | ||
GenBank mRNA | AJ251027.1 | - | 4 - 5 | 427 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180944.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180944.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180944.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900180944.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900180944.hg.1 | TC0900011835.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372032.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000372034.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000461961.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000473737.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | ENST00000618116.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_001288987.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NM_014581.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110243.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011835.hg.1 | NR_110243.1 | Refseq |
