Assay Details
| Gene Symbol: HOPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145460.1 | NP_001138932.1 | 3 - 4 | 733 | 66 | |
| XM_006714052.2 | XP_006714115.1 | 3 - 4 | 728 | 66 | ||
| GenBank mRNA | AF492678.1 | - | 3 - 4 | 358 | 66 | |
| AF492679.1 | - | 3 - 4 | 314 | 66 | ||
| CK002567.1 | - | 2 - 3 | 211 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001222.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
trophectodermal cell differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
heart development
regulation of heart contraction
histone deacetylation
regulation of protein binding
positive regulation of skeletal muscle tissue regeneration
negative regulation of cell differentiation
lung alveolus development
positive regulation of striated muscle cell differentiation
DNA binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
trophectodermal cell differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
heart development
regulation of heart contraction
histone deacetylation
regulation of protein binding
positive regulation of skeletal muscle tissue regeneration
negative regulation of cell differentiation
lung alveolus development
positive regulation of striated muscle cell differentiation
DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000317745.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000317745.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000317745.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000317745.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000317745.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000317745.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000317745.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000317745.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000337881.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381260.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381260.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381260.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381260.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381260.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381260.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381260.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000381260.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000420433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503639.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000503864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000506661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000506661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000508121.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000508121.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000508121.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000508121.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000508121.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000508121.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000508121.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000553379.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000553379.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000553379.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000553379.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000553379.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000553379.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000553379.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000554144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000555760.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000555760.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000555760.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000555760.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000555760.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000555760.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000555760.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000555760.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556376.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556614.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556614.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556614.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556614.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556614.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556614.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556614.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000556614.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000605395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000605395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | ENST00000605395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_001145460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_032495.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139211.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010775.hg.1 | NM_139212.3 | Refseq |

HOPX
Genomic Map