Assay Details
| Gene Symbol: C12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135051.1 | 8 - 9 | 1322 | 152 | ||
| GenBank mRNA | AK097445.1 | - | 8 - 9 | 1271 | 152 | |
| EU668334.1 | - | 8 - 9 | 1271 | 152 | ||
| GQ472216.1 | - | 8 - 9 | 1271 | 152 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATP binding cassette subfamily D member 2 | ABCD2 |
ABC39, ALDL1, ALDR, ALDRP, hALDR |
-32.1kb | Anti-sense | Chr. 12 - 39530966 - 39651333 on NCBI Build GRCh38 |
| Downstream | solute carrier family 2 member 13 | SLC2A13 | HMIT | 70.7kb | Anti-sense | Chr. 12 - 39755021 - 40106085 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000324616.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000405531.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | ENST00000468200.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007349.hg.1 | NM_001031748.3 | Refseq |

C12orf40
Genomic Map