Assay Details
Gene Symbol: IPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK057044.1 | - | 9 - 10 | 1094 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK057044.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
