Assay Details
| Gene Symbol: LOC730098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320037.1 | NP_001306966.1 | 1 - 2 | 80 | 71 | |
| NR_135148.1 | 1 - 2 | 80 | 71 | |||
| GenBank mRNA | DB061900.1 | - | 1 - 2 | 80 | 71 | |
| HY158194.1 | - | 1 - 2 | 75 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320037.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |

LOC730098
Genomic Map