| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004542.hg.1 |
TC04000323.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004542.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004542.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004544.hg.1 |
TC04000323.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004544.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04004544.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007570.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007570.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007570.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007570.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007570.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007570.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007570.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151745.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151745.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151745.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151745.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151745.hg.1 |
TC0400007570.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400151747.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151747.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151747.hg.1 |
ENST00000504250.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400151747.hg.1 |
NR_003935.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400151747.hg.1 |
TC0400007570.hg.1 |
Clariom D TC |