Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017977.hg.1 |
ENST00000291442.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017977.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017977.hg.1 |
TC19001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017979.hg.1 |
ENST00000291442.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017979.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017979.hg.1 |
TC19001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017978.hg.1 |
ENST00000291442.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017978.hg.1 |
ENST00000536940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017978.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017978.hg.1 |
TC19001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017974.hg.1 |
ENST00000291442.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017974.hg.1 |
ENST00000536940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017974.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017974.hg.1 |
TC19001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017976.hg.1 |
ENST00000291442.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017976.hg.1 |
ENST00000536940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017976.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017976.hg.1 |
TC19001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017980.hg.1 |
ENST00000291442.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017980.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017980.hg.1 |
TC19001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017975.hg.1 |
ENST00000291442.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017975.hg.1 |
ENST00000536940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017975.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017975.hg.1 |
TC19001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205112.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205112.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205112.hg.1 |
TC1900011903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900205112.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205112.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205110.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205110.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205110.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205110.hg.1 |
TC1900011903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900205110.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205110.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205110.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205109.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205109.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205109.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205109.hg.1 |
TC1900011903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900205109.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205109.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205109.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205113.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205113.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205113.hg.1 |
TC1900011903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900205113.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205113.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
ENST00000596878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011903.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205114.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205114.hg.1 |
NM_005234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900205114.hg.1 |
TC1900011903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900205114.hg.1 |
ENST00000291442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900205114.hg.1 |
NM_005234.3 |
Refseq |