Assay Details
| Gene Symbol: PNPLA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177675.1 | NP_001171146.1 | 4 - 5 | 739 | 82 | |
| NM_138814.3 | NP_620169.1 | 6 - 7 | 1081 | 82 | ||
| XM_011529959.2 | XP_011528261.1 | 4 - 5 | 566 | 82 | ||
| GenBank mRNA | AK056648.1 | - | 6 - 7 | 1078 | 82 | |
| BC031820.2 | - | 6 - 7 | 1045 | 4778605 | 82 | |
| BC104839.1 | - | 6 - 7 | 995 | 8143842 | 82 | |
| BC104843.1 | - | 6 - 7 | 995 | 8143846 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000865.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22014678.hg.1 | ENST00000216177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014678.hg.1 | ENST00000381198.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014678.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014678.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014678.hg.1 | TC22000865.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008921.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | TC2200008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | NM_138814.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000216177.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000381198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000438734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000593866.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | ENST00000597664.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | NM_001177675.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200164797.hg.1 | NM_138814.3 | Refseq |

PNPLA5
Genomic Map