Assay Details
| Gene Symbol: COX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004376.6 | NP_004367.2 | 8 - 9 | 1716 | 115 | |
| XM_011539298.2 | XP_011537600.1 | 7 - 8 | 1026 | 115 | ||
| GenBank mRNA | AF044323.1 | - | 8 - 9 | 1150 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001584.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial electron transport, cytochrome c to oxygen
heme biosynthetic process
heme a biosynthetic process
respiratory gaseous exchange
respiratory chain complex IV assembly
cellular respiration
oxidation-reduction process
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
oxidoreductase activity, acting on the CH-CH group of donors
oxidoreductase activity, acting on NAD(P)H, heme protein as acceptor
mitochondrial electron transport, cytochrome c to oxygen
heme biosynthetic process
heme a biosynthetic process
respiratory gaseous exchange
respiratory chain complex IV assembly
cellular respiration
oxidation-reduction process
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
oxidoreductase activity, acting on the CH-CH group of donors
oxidoreductase activity, acting on NAD(P)H, heme protein as acceptor
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000497381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000497381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | ENST00000497381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020361.hg.1 | ENST00000370483.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020361.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020361.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020361.hg.1 | TC10001584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020359.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020359.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020359.hg.1 | TC10001584.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000179024.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179024.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179024.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179024.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179024.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179024.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179024.hg.1 | TC1000011592.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000179023.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179023.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179023.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179023.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179023.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179023.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179023.hg.1 | TC1000011592.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000179021.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179021.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179021.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179021.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179021.hg.1 | TC1000011592.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000016171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000370483.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000497381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000497381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | ENST00000497381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_004376.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011592.hg.1 | NM_078470.5 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 123514 | - | - |

COX15
Genomic Map