Assay Details
Gene Symbol: NFKBIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004556.2 | NP_004547.2 | 3 - 4 | 1131 | 77 | |
GenBank mRNA | AK301360.1 | - | 3 - 4 | 895 | 77 | |
BC011676.2 | - | 3 - 4 | 877 | 77 | ||
BC063609.1 | - | 3 - 4 | 877 | 77 | ||
U91616.1 | - | 3 - 4 | 1139 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001762.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06021172.hg.1 | TC06001762.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06021172.hg.1 | ENST00000275015.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06021172.hg.1 | ENST00000477930.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06021172.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000443607.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000443607.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000443607.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000443607.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000443607.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000443607.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011913.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | ENST00000275015.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | ENST00000477930.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | ENST00000616555.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | ENST00000619360.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | NM_004556.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600181924.hg.1 | TC0600011913.hg.1 | Clariom D TC |
