Assay Details
Gene Symbol: WBSCR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022479.2 | NP_071924.1 | 1 - 2 | 502 | 77 | |
XM_011516467.2 | XP_011514769.1 | 1 - 2 | 625 | 77 | ||
XM_011516469.2 | XP_011514771.1 | 1 - 2 | 625 | 77 | ||
XM_017012521.1 | XP_016868010.1 | 1 - 2 | 625 | 77 | ||
GenBank mRNA | AB078148.1 | - | 1 - 2 | 236 | 77 | |
AF410457.1 | - | 1 - 2 | 236 | 77 | ||
AJ626726.1 | - | 1 - 2 | 236 | 77 | ||
AK290036.1 | - | 1 - 2 | 505 | 77 | ||
BC067524.1 | - | 1 - 2 | 236 | 7002169 | 77 | |
BC067525.1 | - | 1 - 2 | 236 | 7002172 | 77 | |
BC069624.1 | - | 1 - 2 | 236 | 7262423 | 77 | |
BC069628.1 | - | 1 - 2 | 236 | 7262435 | 77 | |
BC069636.1 | - | 1 - 2 | 236 | 7262447 | 77 | |
BC069645.1 | - | 1 - 2 | 236 | 7262459 | 77 | |
BC069997.1 | - | 1 - 2 | 236 | 7002171 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000425.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07005710.hg.1 | TC07000425.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07005710.hg.1 | ENST00000333538.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07005710.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07005714.hg.1 | TC07000425.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07005714.hg.1 | ENST00000333538.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07005714.hg.1 | ENST00000498380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07005714.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07005716.hg.1 | TC07000425.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07005716.hg.1 | ENST00000333538.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07005716.hg.1 | ENST00000498380.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07005716.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700154678.hg.1 | TC0700007925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700154672.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154672.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700154672.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154672.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700154672.hg.1 | TC0700007925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000447516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000467723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000498380.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | ENST00000618959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007925.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700154673.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154673.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700154673.hg.1 | ENST00000333538.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700154673.hg.1 | NM_022479.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700154673.hg.1 | TC0700007925.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 175815 | - | - |
