Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008084.hg.1 |
TC0X000616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008084.hg.1 |
ENST00000357121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008084.hg.1 |
ENST00000371113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008084.hg.1 |
NM_000276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008084.hg.1 |
NM_001587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
ENST00000357121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
ENST00000371113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
NM_000276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
NM_001587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
ENST00000357121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
ENST00000371113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
NM_000276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
NM_001587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157052.hg.1 |
TC0X00008381.hg.1 |
Clariom D TC |