| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14014357.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14014357.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14014357.hg.1 |
TC14001263.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400170498.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400170498.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400170498.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400170498.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400170498.hg.1 |
TC1400009563.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
ENST00000216540.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009563.hg.1 |
NM_003049.3 |
Refseq |