Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043255.hg.1 |
TC01002769.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043255.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043255.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014562.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208701.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208701.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208701.hg.1 |
ENST00000370952.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100208701.hg.1 |
NM_017768.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100208701.hg.1 |
TC0100014562.hg.1 |
Clariom D TC |