Assay Details
Gene Symbol: DDX43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018665.2 | NP_061135.2 | 1 - 2 | 597 | 146 | |
XM_011535926.1 | XP_011534228.1 | 1 - 2 | 597 | 146 | ||
XM_017011003.1 | XP_016866492.1 | 1 - 2 | 265 | 146 | ||
GenBank mRNA | AJ278110.1 | - | 1 - 2 | 460 | 146 | |
AK303324.1 | - | 1 - 2 | 306 | 146 | ||
AK313686.1 | - | 1 - 2 | 297 | 146 | ||
AL136751.1 | - | 1 - 2 | 285 | 146 | ||
BC066938.1 | - | 1 - 2 | 278 | 4829072 | 146 | |
BX646425.1 | - | 1 - 2 | 344 | 146 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000733.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017011003.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06008755.hg.1 | TC06000733.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06008755.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06008755.hg.1 | ENST00000539829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06008755.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06008756.hg.1 | TC06000733.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06008756.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06008756.hg.1 | ENST00000539829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06008756.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06008753.hg.1 | TC06000733.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06008753.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06008753.hg.1 | ENST00000539829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06008753.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06008751.hg.1 | TC06000733.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06008751.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370359.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011641.hg.1 | ENST00000441064.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06008752.hg.1 | TC06000733.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06008752.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06008752.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011764.hg.1 | ENST00000510713.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159252.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159252.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159252.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159252.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159252.hg.1 | TC0600008527.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500011764.hg.1 | ENST00000510713.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370359.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000370363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | ENST00000441145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012289.hg.1 | NM_001080507.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159251.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159251.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159251.hg.1 | TC0600008527.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600159256.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159256.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159256.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159256.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159256.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159256.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159256.hg.1 | TC0600008527.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | ENST00000479773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008527.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011641.hg.1 | ENST00000441064.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159255.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159255.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159255.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159255.hg.1 | ENST00000370336.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159255.hg.1 | ENST00000464221.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600159255.hg.1 | NM_018665.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600159255.hg.1 | TC0600008527.hg.1 | Clariom D TC |
