Assay Details
| Gene Symbol: ZPBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199321.2 | NP_955353.1 | 1 - 2 | 209 | 112 | |
| XM_011524298.1 | XP_011522600.1 | 1 - 2 | 247 | 112 | ||
| GenBank mRNA | AK292383.1 | - | 1 - 2 | 247 | 112 | |
| AY251603.1 | - | 1 - 2 | 192 | 112 | ||
| DB056422.1 | - | 1 - 2 | 247 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000482.hg
Additional Information:
This assay is in the 5'UTR region of the AK292383.1,AY251603.1,NM_199321.2,XM_011524298.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005744.hg.1 | ENST00000348931.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005744.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005744.hg.1 | TC17000482.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156310.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156310.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156310.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156310.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156310.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156310.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156310.hg.1 | TC1700007761.hg.1 | Clariom D TC |

ZPBP2
Genomic Map