| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027680.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027680.hg.1 |
ENST00000424261.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027680.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027680.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027680.hg.1 |
TC11002314.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027677.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027677.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027677.hg.1 |
TC11002314.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027678.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027678.hg.1 |
ENST00000424261.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027678.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027678.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027678.hg.1 |
TC11002314.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027679.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027679.hg.1 |
ENST00000424261.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027679.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027679.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11027679.hg.1 |
TC11002314.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012382.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195110.hg.1 |
TC1100012382.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195112.hg.1 |
TC1100012382.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
ENST00000424261.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
NM_017678.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195109.hg.1 |
TC1100012382.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100195108.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195108.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195108.hg.1 |
ENST00000375478.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100195108.hg.1 |
NM_001077639.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100195108.hg.1 |
TC1100012382.hg.1 |
Clariom D TC |