Assay Details
Gene Symbol: ZDHHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001323627.1 | NP_001310556.1 | 1 - 2 | 319 | 107 | |
NM_013304.2 | NP_037436.1 | 1 - 2 | 261 | 107 | ||
GenBank mRNA | BC021908.1 | - | 1 - 2 | 287 | 4053374 | 107 |
HY054038.1 | - | 1 - 2 | 320 | 107 | ||
U90653.2 | - | 1 - 2 | 261 | 107 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001189.hg
Additional Information:
This assay is in the 5'UTR region of the BC021908.1,NM_001323627.1,NM_013304.2,U90653.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16016541.hg.1 | ENST00000348579.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16016541.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016541.hg.1 | TC16001189.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16016540.hg.1 | ENST00000348579.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16016540.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016540.hg.1 | TC16001189.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016543.hg.1 | ENST00000348579.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16016543.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016543.hg.1 | TC16001189.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600181935.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181935.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181935.hg.1 | TC1600010628.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600181933.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181933.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181933.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600181933.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181933.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181933.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600181933.hg.1 | TC1600010628.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600181934.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181934.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600181934.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181934.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600181934.hg.1 | TC1600010628.hg.1 | Clariom D TC |
