Assay Details
Gene Symbol: ZDHHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001323627.1 | NP_001310556.1 | 7 - 8 | 1178 | 67 | |
NM_013304.2 | NP_037436.1 | 7 - 8 | 1120 | 67 | ||
XM_005255915.1 | XP_005255972.1 | 6 - 7 | 854 | 67 | ||
XM_006721185.1 | XP_006721248.1 | 6 - 7 | 854 | 67 | ||
XM_011523058.1 | XP_011521360.1 | 6 - 7 | 854 | 67 | ||
XM_011523059.1 | XP_011521361.1 | 6 - 7 | 854 | 67 | ||
XM_011523060.1 | XP_011521362.1 | 5 - 6 | 653 | 67 | ||
GenBank mRNA | BC021908.1 | - | 7 - 8 | 1146 | 4053374 | 67 |
U90653.2 | - | 7 - 8 | 1120 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001189.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016534.hg.1 | ENST00000348579.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16016534.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016534.hg.1 | TC16001189.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000566075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000567311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | ENST00000568650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010628.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | ENST00000348579.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | ENST00000562122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | ENST00000565726.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | NM_013304.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600181921.hg.1 | TC1600010628.hg.1 | Clariom D TC |
