Assay Details
| Gene Symbol: MBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204140.1 | NP_001191069.1 | 11 - 12 | 1491 | 86 | |
| NM_001204143.1 | NP_001191072.1 | 10 - 11 | 1197 | 86 | ||
| NM_001323952.1 | NP_001310881.1 | 10 - 11 | 1341 | 86 | ||
| NM_002384.2 | NP_002375.1 | 10 - 11 | 1416 | 86 | ||
| NM_015844.2 | NP_056669.2 | 10 - 11 | 1416 | 86 | ||
| XM_011526001.1 | XP_011524303.1 | 10 - 11 | 1163 | 86 | ||
| XM_011526006.1 | XP_011524308.1 | 10 - 11 | 1163 | 86 | ||
| XM_017025765.1 | XP_016881254.1 | 10 - 11 | 1163 | 86 | ||
| XM_017025766.1 | XP_016881255.1 | 11 - 12 | 1238 | 86 | ||
| XM_017025767.1 | XP_016881256.1 | 11 - 12 | 1238 | 86 | ||
| XM_017025768.1 | XP_016881257.1 | 10 - 11 | 1163 | 86 | ||
| XM_017025770.1 | XP_016881259.1 | 10 - 11 | 1163 | 86 | ||
| XM_017025771.1 | XP_016881260.1 | 10 - 11 | 1163 | 86 | ||
| XM_017025772.1 | XP_016881261.1 | 10 - 11 | 1163 | 86 | ||
| XM_017025775.1 | XP_016881264.1 | 10 - 11 | 1163 | 86 | ||
| XM_017025776.1 | XP_016881265.1 | 10 - 11 | 1163 | 86 | ||
| GenBank mRNA | AB209765.1 | - | 7 - 8 | 681 | 86 | |
| AF078833.1 | - | 9 - 10 | 979 | 86 | ||
| AK300759.1 | - | 10 - 11 | 1197 | 86 | ||
| BC033242.1 | - | 10 - 11 | 1142 | 5432943 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

MBD1
Genomic Map