Assay Details
| Gene Symbol: FRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006653.4 | NP_006644.1 | 2 - 3 | 236 | 121 | |
| XM_011514254.2 | XP_011512556.1 | 2 - 3 | 1089 | 121 | ||
| XM_011514255.2 | XP_011512557.1 | 2 - 3 | 1084 | 121 | ||
| XM_017010193.1 | XP_016865682.1 | 1 - 2 | 1642 | 121 | ||
| GenBank mRNA | HY036161.1 | - | 2 - 3 | 237 | 121 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001723.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_006653.4,XM_011514254.2,XM_011514255.2,XM_017010193.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
signal transduction
fibroblast growth factor receptor signaling pathway
positive regulation of GTPase activity
Ras guanyl-nucleotide exchange factor activity
fibroblast growth factor receptor binding
insulin receptor binding
protein binding
identical protein binding
MAPK cascade
signal transduction
fibroblast growth factor receptor signaling pathway
positive regulation of GTPase activity
Ras guanyl-nucleotide exchange factor activity
fibroblast growth factor receptor binding
insulin receptor binding
protein binding
identical protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06020550.hg.1 | TC06001723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020550.hg.1 | ENST00000259748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020550.hg.1 | ENST00000373018.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020550.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020550.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020552.hg.1 | TC06001723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020552.hg.1 | ENST00000373018.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020552.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020547.hg.1 | TC06001723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020547.hg.1 | ENST00000259748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020547.hg.1 | ENST00000373018.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020547.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020547.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020549.hg.1 | TC06001723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020549.hg.1 | ENST00000259748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020549.hg.1 | ENST00000373018.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020549.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06020549.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181040.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181040.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181040.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181040.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181040.hg.1 | TC0600011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181034.hg.1 | TC0600011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181038.hg.1 | TC0600011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181037.hg.1 | TC0600011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000259748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000373018.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000422888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000426290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | ENST00000466420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011802.hg.1 | NM_006653.4 | Refseq |

FRS3
Genomic Map