Assay Details
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016037.3 | NP_057121.2 | 1 - 2 | 149 | 56 | |
GenBank mRNA | AK291900.1 | - | 1 - 2 | 79 | 56 | |
AK296018.1 | - | 1 - 2 | 82 | 56 | ||
BC005182.1 | - | 1 - 2 | 94 | 3995984 | 56 | |
BU940926.1 | - | 1 - 2 | 152 | 56 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000484.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01007461.hg.1 | TC01000484.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01007461.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007461.hg.1 | ENST00000537711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007461.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01007462.hg.1 | TC01000484.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01007462.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007462.hg.1 | ENST00000537711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007462.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155318.hg.1 | TC0100007858.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | ENST00000488453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007858.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155315.hg.1 | TC0100007858.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155316.hg.1 | TC0100007858.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | ENST00000373014.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | ENST00000483182.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | ENST00000486563.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | NM_016037.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100155317.hg.1 | TC0100007858.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 159208 | - | - |
