Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013873.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013873.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013873.hg.1 |
TC20001010.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013872.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013872.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013872.hg.1 |
TC20001010.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000168020.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168020.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168020.hg.1 |
TC2000009751.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168020.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168020.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168021.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168021.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168021.hg.1 |
TC2000009751.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168021.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168021.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
ENST00000252998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009751.hg.1 |
NM_080833.2 |
Refseq |