Assay Details
| Gene Symbol: SRRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194286.3 | NP_919262.2 | 12 - 13 | 1926 | 62 | |
| GenBank mRNA | AB058756.1 | - | 12 - 13 | 1830 | 62 | |
| AK291361.1 | - | 12 - 13 | 1926 | 62 | ||
| BC152471.1 | - | 12 - 13 | 1841 | 8862608 | 62 | |
| BC167162.1 | - | 12 - 13 | 1830 | 8862617 | 62 | |
| BC167163.1 | - | 12 - 13 | 1830 | 8862618 | 62 | |
| KC995185.1 | - | 12 - 13 | 1538 | 62 | ||
| KC995186.1 | - | 12 - 13 | 1538 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000930.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12012082.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012082.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012082.hg.1 | TC12000930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012083.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012083.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012083.hg.1 | TC12000930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012085.hg.1 | TC12000930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167163.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167163.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167163.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167163.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167163.hg.1 | TC1200009085.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200167159.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167159.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167159.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167159.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167159.hg.1 | TC1200009085.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000537597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | ENST00000545224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009085.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167162.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167162.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167162.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167162.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167162.hg.1 | TC1200009085.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200167160.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167160.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167160.hg.1 | ENST00000267260.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167160.hg.1 | NM_194286.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167160.hg.1 | TC1200009085.hg.1 | Clariom D TC |

SRRM4
Genomic Map