Assay Details
Gene Symbol: ACADS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000017.3 | NP_000008.1 | 6 - 7 | 943 | 52 | |
NM_001302554.1 | NP_001289483.1 | 6 - 7 | 931 | 52 | ||
GenBank mRNA | AB527081.1 | - | 6 - 7 | 795 | 52 | |
AK300645.1 | - | 6 - 7 | 809 | 52 | ||
BC025963.1 | - | 6 - 7 | 842 | 4842286 | 52 | |
M26393.1 | - | 6 - 7 | 827 | 52 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000951.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to starvation
butyrate catabolic process
protein homotetramerization
response to glucocorticoid
lipid homeostasis
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
butyryl-CoA dehydrogenase activity
electron carrier activity
flavin adenine dinucleotide binding
fatty acid beta-oxidation
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to starvation
butyrate catabolic process
protein homotetramerization
response to glucocorticoid
lipid homeostasis
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
butyryl-CoA dehydrogenase activity
electron carrier activity
flavin adenine dinucleotide binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12012354.hg.1 | ENST00000242592.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012354.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012354.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12012354.hg.1 | TC12000951.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | ENST00000242592.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | ENST00000411593.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | NM_000017.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | NM_001302554.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200167631.hg.1 | TC1200009141.hg.1 | Clariom D TC |
