Assay Details
| Gene Symbol: ACADS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000017.3 | NP_000008.1 | 7 - 8 | 1078 | 94 | |
| NM_001302554.1 | NP_001289483.1 | 7 - 8 | 1066 | 94 | ||
| GenBank mRNA | AB527081.1 | - | 7 - 8 | 930 | 94 | |
| BC025963.1 | - | 7 - 8 | 977 | 4842286 | 94 | |
| M26393.1 | - | 7 - 8 | 962 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000951.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to starvation
butyrate catabolic process
protein homotetramerization
response to glucocorticoid
lipid homeostasis
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
butyryl-CoA dehydrogenase activity
electron carrier activity
flavin adenine dinucleotide binding
fatty acid beta-oxidation
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to starvation
butyrate catabolic process
protein homotetramerization
response to glucocorticoid
lipid homeostasis
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
butyryl-CoA dehydrogenase activity
electron carrier activity
flavin adenine dinucleotide binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12012355.hg.1 | ENST00000242592.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012355.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012355.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012355.hg.1 | TC12000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012356.hg.1 | ENST00000242592.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012356.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012356.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012356.hg.1 | TC12000951.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167632.hg.1 | TC1200009141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000242592.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000411593.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | ENST00000539690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_000017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009141.hg.1 | NM_001302554.1 | Refseq |

ACADS
Genomic Map