Assay Details
Gene Symbol: C11orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276722.1 | NP_001263651.1 | 4 - 5 | 652 | 77 | |
NM_001276723.1 | NP_001263652.1 | 4 - 5 | 532 | 77 | ||
NM_001276724.1 | NP_001263653.1 | 4 - 5 | 527 | 77 | ||
NM_138787.3 | NP_620142.2 | 4 - 5 | 483 | 77 | ||
XM_006718138.3 | XP_006718201.1 | 5 - 6 | 553 | 77 | ||
XM_011519885.2 | XP_011518187.1 | 5 - 6 | 818 | 77 | ||
GenBank mRNA | AK290833.1 | - | 4 - 5 | 476 | 77 | |
AK299642.1 | - | 4 - 5 | 459 | 77 | ||
BC051031.1 | - | 4 - 5 | 433 | 5122890 | 77 | |
BI831382.1 | - | 4 - 5 | 444 | 77 | ||
CD678640.1 | - | 4 - 5 | 438 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
