Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061095.hg.1 |
TC01004046.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061095.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061095.hg.1 |
ENST00000449326.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061095.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000417765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000417765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000417765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000417765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235625.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235625.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235625.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100235625.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100235625.hg.1 |
TC0100018006.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000366535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000417765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000417765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000417765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000417765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000462358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
ENST00000468215.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018006.hg.1 |
NM_001126.3 |
Refseq |