Assay Details
Gene Symbol: TPD52L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003396.2 | NP_001003396.1 | 4 - 5 | 604 | 78 | |
XM_017011239.1 | XP_016866728.1 | 4 - 5 | 706 | 78 | ||
GenBank mRNA | BC002375.2 | - | 4 - 5 | 494 | 2822877 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
