Assay Details
| Gene Symbol: RNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018683.3 | NP_061153.1 | 2 - 3 | 325 | 72 | |
| GenBank mRNA | AF265215.1 | - | 2 - 3 | 295 | 72 | |
| AK091976.1 | - | 2 - 3 | 303 | 72 | ||
| AK301731.1 | - | 2 - 3 | 295 | 72 | ||
| AK315638.1 | - | 2 - 3 | 305 | 72 | ||
| BC013695.1 | - | 2 - 3 | 294 | 3858611 | 72 | |
| BC066919.1 | - | 2 - 3 | 303 | 5264729 | 72 | |
| BM830132.1 | - | 2 - 3 | 323 | 72 | ||
| BT006795.1 | - | 2 - 3 | 289 | 72 | ||
| BX640603.1 | - | 2 - 3 | 307 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000393.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin conjugating enzyme binding
ubiquitin protein ligase activity
protein polyubiquitination
multicellular organism development
spermatogenesis
cell differentiation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin conjugating enzyme binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20005674.hg.1 | ENST00000244061.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005674.hg.1 | ENST00000449816.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005674.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20005674.hg.1 | TC20000393.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | TC2000007666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000153470.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000244061.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000622999.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000623732.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000624666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | ENST00000625177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007666.hg.1 | NM_018683.3 | Refseq |

RNF114
Genomic Map