| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005676.hg.1 |
ENST00000244061.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005676.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005676.hg.1 |
TC20000393.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
TC2000007666.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153474.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000244061.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622920.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000622999.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623528.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000623732.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000624666.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
ENST00000625177.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007666.hg.1 |
NM_018683.3 |
Refseq |