Assay Details
| Gene Symbol: SNRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003092.4 | NP_003083.1 | 4 - 5 | 618 | 87 | |
| NM_198220.2 | NP_937863.1 | 4 - 5 | 518 | 87 | ||
| GenBank mRNA | AB451234.1 | - | 3 - 4 | 382 | 87 | |
| AB451359.1 | - | 3 - 4 | 382 | 87 | ||
| AK313004.1 | - | 4 - 5 | 552 | 87 | ||
| BC018022.1 | - | 4 - 5 | 570 | 4286926 | 87 | |
| BC036737.2 | - | 4 - 5 | 477 | 5533477 | 87 | |
| DA088017.1 | - | 4 - 5 | 518 | 87 | ||
| M15841.1 | - | 4 - 5 | 507 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000104.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20001406.hg.1 | ENST00000246071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001406.hg.1 | ENST00000377943.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001406.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001406.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001406.hg.1 | TC20000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011906.hg.1 | ENST00000412744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001405.hg.1 | ENST00000246071.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001405.hg.1 | ENST00000377943.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001405.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001405.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20001405.hg.1 | TC20000104.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146444.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146445.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011906.hg.1 | ENST00000412744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | ENST00000478522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006781.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | NM_198220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | TC2000006781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | ENST00000246071.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | ENST00000377943.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | NM_003092.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000146443.hg.1 | NM_198220.2 | Refseq |

SNRPB2
Genomic Map