Assay Details
| Gene Symbol: NDUFS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BG706812.1 | - | 4 - 5 | 540 | 95 | |
| BM172493.1 | - | 4 - 5 | 405 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFS6| Gene Symbol: NDUFS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BG706812.1 | - | 4 - 5 | 540 | 95 | |
| BM172493.1 | - | 4 - 5 | 405 | 94 | ||
Set Membership: |
|