Assay Details
Gene Symbol: EXOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007277.4 | NP_009208.2 | 2 - 3 | 275 | 72 | |
XM_011513948.2 | XP_011512250.1 | 2 - 3 | 310 | 72 | ||
XM_011513949.2 | XP_011512251.1 | 2 - 3 | 327 | 72 | ||
XM_011513950.2 | XP_011512252.1 | 1 - 2 | 205 | 72 | ||
XM_011513951.2 | XP_011512253.1 | 2 - 3 | 304 | 72 | ||
XM_017009000.1 | XP_016864489.1 | 2 - 3 | 310 | 72 | ||
XM_017009001.1 | XP_016864490.1 | 2 - 3 | 310 | 72 | ||
XM_017009002.1 | XP_016864491.1 | 2 - 3 | 310 | 72 | ||
XM_017009003.1 | XP_016864492.1 | 2 - 3 | 310 | 72 | ||
XM_017009004.1 | XP_016864493.1 | 3 - 4 | 388 | 72 | ||
XM_017009005.1 | XP_016864494.1 | 2 - 3 | 304 | 72 | ||
XM_017009006.1 | XP_016864495.1 | 2 - 3 | 304 | 72 | ||
GenBank mRNA | AK074086.1 | - | 1 - 2 | 266 | 72 | |
AL545132.3 | - | 2 - 3 | 308 | 72 | ||
AL589499.1 | - | 289 | 72 | |||
BC064569.1 | - | 1 - 2 | 217 | 5590332 | 72 | |
GU727644.1 | - | 2 - 3 | 275 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000006.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05000176.hg.1 | TC05000006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05000176.hg.1 | ENST00000315013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000176.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000176.hg.1 | ENST00000512944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000176.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05000179.hg.1 | TC05000006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05000179.hg.1 | ENST00000315013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000179.hg.1 | ENST00000340158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000179.hg.1 | ENST00000512944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000179.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05000177.hg.1 | TC05000006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05000177.hg.1 | ENST00000315013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000177.hg.1 | ENST00000340158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000177.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000177.hg.1 | ENST00000512944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000177.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05000180.hg.1 | TC05000006.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05000180.hg.1 | ENST00000315013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000180.hg.1 | ENST00000340158.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000180.hg.1 | ENST00000512944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05000180.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000505947.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000511015.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013142.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192566.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192566.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192566.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192566.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192566.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192566.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192566.hg.1 | TC0500013142.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500192575.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192575.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192575.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192575.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192575.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192575.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192575.hg.1 | TC0500013142.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192571.hg.1 | TC0500013142.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | ENST00000510441.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192572.hg.1 | TC0500013142.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | ENST00000315013.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | ENST00000508022.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | ENST00000512944.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | NM_007277.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500192574.hg.1 | TC0500013142.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 173982 | - | - |
