Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000505947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000505947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000505947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000505947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000508022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000508022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000508022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000508022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000183.hg.1 |
TC05000006.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000183.hg.1 |
ENST00000315013.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000183.hg.1 |
ENST00000512944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05000183.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192577.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192577.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192577.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192577.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192577.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500192577.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500192577.hg.1 |
TC0500013142.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000315013.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000505947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000505947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000505947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000505947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000508022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000508022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000508022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000508022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000510441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000511015.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
ENST00000512944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013142.hg.1 |
NM_007277.4 |
Refseq |