Assay Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035505.1 | NP_001030582.1 | 1 - 2 | 94 | 88 | |
NM_212552.2 | NP_997717.2 | 1 - 2 | 94 | 88 | ||
GenBank mRNA | BC042036.1 | - | 1 - 2 | 90 | 88 | |
BU600037.1 | - | 99 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001992.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001035505.1,NM_212552.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02031719.hg.1 | TC02001992.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02031719.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02031719.hg.1 | ENST00000327428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02031719.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02031719.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02031720.hg.1 | TC02001992.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02031720.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02031720.hg.1 | ENST00000327428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02031720.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02031720.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02031714.hg.1 | TC02001992.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02031714.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02031714.hg.1 | ENST00000327428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02031714.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02031714.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006920.hg.1 | ENST00000586496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006920.hg.1 | ENST00000586496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011927.hg.1 | ENST00000505182.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195575.hg.1 | TC0200013095.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006920.hg.1 | ENST00000586496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006920.hg.1 | ENST00000586496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195580.hg.1 | TC0200013095.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195582.hg.1 | TC0200013095.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500011927.hg.1 | ENST00000505182.1 | Ensembl |
