Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006920.hg.1 |
ENST00000586496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006920.hg.1 |
ENST00000586496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031713.hg.1 |
TC02001992.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031713.hg.1 |
ENST00000327428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031713.hg.1 |
NM_212552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006920.hg.1 |
ENST00000586496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006920.hg.1 |
ENST00000586496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195574.hg.1 |
ENST00000327428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195574.hg.1 |
ENST00000477685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195574.hg.1 |
NM_212552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195574.hg.1 |
ENST00000327428.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195574.hg.1 |
ENST00000477685.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195574.hg.1 |
NM_212552.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195574.hg.1 |
TC0200013095.hg.1 |
Clariom D TC |