Assay Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212552.2 | NP_997717.2 | 3 - 4 | 301 | 63 | |
GenBank mRNA | AI652095.1 | - | 1 - 2 | 63 | ||
BC017744.2 | - | 2 | 137 | 63 | ||
BC042036.1 | - | 3 - 4 | 297 | 63 | ||
BU600037.1 | - | 306 | 63 | |||
DQ225187.1 | - | 3 - 4 | 263 | 63 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001992.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02031712.hg.1 | TC02001992.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02031712.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02031712.hg.1 | ENST00000327428.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02031712.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02031712.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200195573.hg.1 | TC0200013095.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000295326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000327428.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000469676.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000477685.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | ENST00000484655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_001035505.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013095.hg.1 | NM_212552.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 172828 | - | - |
