Assay Details
| Gene Symbol: IKZF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257409.1 | NP_001244338.1 | 4 - 5 | 557 | 102 | |
| NM_183229.2 | NP_899052.1 | 5 - 6 | 659 | 102 | ||
| GenBank mRNA | AJ292566.1 | - | 4 - 5 | 597 | 102 | |
| AY377975.1 | - | 4 - 5 | 557 | 102 | ||
More Information
Set Membership: |
|

IKZF3
Genomic Map