| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
TC2100008467.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000291691.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000397680.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000397682.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000445935.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000475776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
NM_001286476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
NM_001286477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
NM_058180.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000291691.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000397680.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000397682.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000445935.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
ENST00000475776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
NM_001286476.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
NM_001286477.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100157488.hg.1 |
NM_058180.4 |
Refseq |